Learn More
Zirconium dichloride oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds an application as rubber additive and fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Melting Point | 400°C (decomposition) |
| Physical Form | Lump |
| Quantity | 50 g |
| Assay Percent Range | 99.9%, metal basis |
| Solubility Information | Soluble in water and alcohol. |
| IUPAC Name | oxozirconium;octahydrate;dihydrochloride |
| Formula Weight | 178.13 (Anhydrous) |
| Sensitivity | Hygroscopic |
| Density | 1.91 |
| Chemical Name or Material | Zirconium Dichloride Oxide |
Sécurité et traitement
- Zirconium dichloride oxide hydrate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Zirconyl chloride hydrate
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.