missing translation for 'onlineSavingsMsg'
Learn More

Zirconium dinitrate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11319298
Click to view available options
Quantity:
25 g
100 g
500 g

Used as a precursor in the production of Pb(Zr0.50Ti0.50)O3 ferroelectric thin films and silicaand alumina-doped nanocrystalline ceria/zirconia. It is also used to prepare cerium oxide/zirconium oxide mixed oxide catalyst supports.

Zirconium dinitrate oxide hydrate is used as a precursor in the production of Pb(Zr0.50Ti0.50)O3 ferroelectric thin films and silicaand alumina-doped nanocrystalline ceria/zirconia. It is also used to prepare cerium oxide/zirconium oxide mixed oxide catalyst supports.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14985-18-3
Molecular Formula N2O7Zr
Molecular Weight (g/mol) 231.23
MDL Number MFCD00149896
InChI Key JWBWFTANBHYHBC-UHFFFAOYSA-N
Synonym zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis
PubChem CID 16211674
SMILES [O-][N+](=O)O[Zr](=O)O[N+]([O-])=O

Specifications

Physical Form Crystalline
Quantity 500 g
UN Number UN1477
Solubility Information Soluble in alcohol.
IUPAC Name (nitrooxy)(oxo)zirconio nitrate
Formula Weight 231.23 (anhydrous)
Percent Purity 99.9%
Packaging Plastic bottle
Chemical Name or Material Zirconium dinitrate oxide hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'