missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium dinitrate oxide hydrate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Zirconium dintrate oxide hydrate is used to prepare CeO2-ZrO2 mixed oxide catalyst supports. It may be used as zirconium precursor in the preparation of Pb(Zr0.50Ti0.50)O3 ferroelectric thin films and Silica- and alumina-doped nanocrystalline ceria/zirconia (Ce0.5Zr0.5O2).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
Physical Form | Powder |
Quantity | 5 g |
Linear Formula | ZrO(NO3)2·xH2O |
UN Number | UN1477 |
Solubility Information | Soluble in alcohol |
IUPAC Name | (nitrooxy)(oxo)zirconio nitrate |
Formula Weight | 231.23 (Anhydrous) |
Percent Purity | 99.994% |
Grade | Puratronic™ |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'