missing translation for 'onlineSavingsMsg'
Learn More

Zirconium dinitrate oxide hydrate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals

Product Code. 11382058
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium dintrate oxide hydrate is used to prepare CeO2-ZrO2 mixed oxide catalyst supports. It may be used as zirconium precursor in the preparation of Pb(Zr0.50Ti0.50)O3 ferroelectric thin films and Silica- and alumina-doped nanocrystalline ceria/zirconia (Ce0.5Zr0.5O2).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14985-18-3
Molecular Formula N2O7Zr
Molecular Weight (g/mol) 231.23
MDL Number MFCD00149896
InChI Key JWBWFTANBHYHBC-UHFFFAOYSA-N
Synonym zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis
PubChem CID 16211674
SMILES [O-][N+](=O)O[Zr](=O)O[N+]([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 100 g
Linear Formula ZrO(NO3)2·xH2O
UN Number UN1477
Solubility Information Soluble in alcohol
IUPAC Name (nitrooxy)(oxo)zirconio nitrate
Formula Weight 231.23 (Anhydrous)
Percent Purity 99.994%
Grade Puratronic™
Odor Odorless
Assay (metals basis)
Chemical Name or Material Zirconium dinitrate oxide hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'