missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) bromide, 99%, Thermo Scientific Chemicals

109.00 € - 527.00 €

Chemical Identifiers

CAS 13777-25-8
Molecular Formula Br4Zr
Molecular Weight (g/mol) 410.84
MDL Number MFCD00049637
InChI Key LSWWNKUULMMMIL-UHFFFAOYSA-J
Synonym zirconium tetrabromide, zrbr4, zirconium iv bromide, zirconium bromide zrbr4 , t-4, zirconium bromide zrbr4, zirconium bromide zrbr4 8ci 9ci, zirconium iv bromide, anhydrous trace metals basis
PubChem CID 83727
IUPAC Name tetrabromozirconium
SMILES [Br-].[Br-].[Br-].[Br-].[Zr+4]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11324949
View Documents Promotion Details
Thermo Scientific Alfa Aesar
086139.09
10 g
109.00 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11334949
View Documents Promotion Details
Thermo Scientific Alfa Aesar
086139.18
50 g
527.00 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zirconium(IV) bromide is used as a starting material in the synthesis of zirconium hydride complexes as well as unique cluster compounds. It is also used as a biochemical for proteomics research and as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13777-25-8
410.84
LSWWNKUULMMMIL-UHFFFAOYSA-J
83727
[Br-].[Br-].[Br-].[Br-].[Zr+4]
Br4Zr
MFCD00049637
zirconium tetrabromide, zrbr4, zirconium iv bromide, zirconium bromide zrbr4 , t-4, zirconium bromide zrbr4, zirconium bromide zrbr4 8ci 9ci, zirconium iv bromide, anhydrous trace metals basis
tetrabromozirconium

Specifications

450° (15 atm)
Powder
Br4Zr
zirconium tetrabromide, zrbr4, zirconium iv bromide, zirconium bromide zrbr4 , t-4, zirconium bromide zrbr4, zirconium bromide zrbr4 8ci 9ci, zirconium iv bromide, anhydrous trace metals basis
LSWWNKUULMMMIL-UHFFFAOYSA-J
tetrabromozirconium
83727
Moisture sensitive
Zirconium(IV) Bromide
13777-25-8
99%
MFCD00049637
Soluble in water.
[Br-].[Br-].[Br-].[Br-].[Zr+4]
410.84
410.86
4.201
Videos
Safety and Handling

Safety and Handling

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : DOT Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 237-417-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only