missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) chloride, 98%, anhydrous

Product Code. p-40907 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10114980

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10026-11-6 | Cl4Zr | 233.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-11-6
Molecular Formula Cl4Zr
Molecular Weight (g/mol) 233.03
MDL Number MFCD00011306
Synonym Zirconium tetrachloride

Specifications

Melting Point 437.0°C
Color White
Physical Form Fine Powder
Quantity 5 g
Assay Percent Range 98%
Linear Formula ZrCl4
Beilstein 01,1295
Fieser 13,16
Merck Index 15,10374
Solubility Information Solubility in water: reacts. Other solubilities: forming hcl and zro2, soluble in alcohol and ether
Formula Weight 233.03
Percent Purity 98%
Packaging Glass bottle
Density 2.8
Chemical Name or Material Zirconium(IV) chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Zirconium(IV) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.