missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) fluoride, 98%, Thermo Scientific Chemicals

Product Code. p-7080354
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11394939

Brand: Thermo Scientific Alfa Aesar 086112.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zirconium(IV) fluoride is used as a zirconium source in oxygen-sensitive applications such as metal production. Together with uranium salt, zirconium fluoride can be a component of fuel-coolant in molten salt reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-64-4
Molecular Formula F4Zr
MDL Number MFCD00011307
Synonym Zirconium tetrafluoride

Specifications

Melting Point 600°C (sublimation)
Color White
Physical Form Powder
Quantity 50 g
Linear Formula ZrF4
UN Number UN3260
Merck Index 14,10175
Solubility Information Slightly soluble in water; Soluble in hydrofluoric acid
Formula Weight 167.21
Percent Purity 98%
Refractive Index 1.59
Sensitivity Moisture sensitive
Density 4.43 g/mL
Chemical Name or Material Zirconium (IV) fluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.