missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) fluoride, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11368288
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a zirconium source in oxygen-sensitive applications, e.g. metal production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-64-4
MDL Number MFCD00011307

Specifications

Melting Point 600°C (sublimation)
Color White
Physical Form Sintered Pieces
Quantity 25 g
Linear Formula ZrF4
UN Number UN3260
Merck Index 14,10175
Solubility Information Soluble in water (13.2g/L @ 20 C)
Formula Weight 167.21
Percent Purity 99.9%
Refractive Index 1.59
Sensitivity Moisture sensitive
Density 4.43 g/mL
Assay (metals basis)
Chemical Name or Material Zirconium(IV) fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'