Learn More
Zirconium(IV) isopropoxide isopropanol complex, Thermo Scientific Chemicals
132.00 € - 546.00 €
Chemical Identifiers
CAS | 14717-56-7 |
---|---|
Molecular Formula | C15H36O5Zr |
Molecular Weight (g/mol) | 387.67 |
MDL Number | MFCD00075157,MFCD00078045 |
InChI Key | NIJDLRJGRCHJDB-UHFFFAOYSA-N |
Synonym | isopropanoltetrakis isopropanolato zirconium |
PubChem CID | 131848446 |
IUPAC Name | propan-2-ol;propan-2-olate;zirconium(4+) |
SMILES | [Zr+4].CC(C)O.CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11304949
|
Thermo Scientific Alfa Aesar
086118.09 |
10 g |
132.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314949
|
Thermo Scientific Alfa Aesar
086118.18 |
50 g |
546.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zirconium(IV) isopropoxide isopropanol is a suitable precursor for the synthesis of olefin-functionalized zirconium containing monomers. It is used in the manufacture of hybrid zirconium containing polymers. It is also used as a source of zirconia, a well known material having high mechanical strength, chemical durability and alkali resistance. It is also used for the chemical protection of metal substrates from acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14717-56-7 | |
387.67 | |
NIJDLRJGRCHJDB-UHFFFAOYSA-N | |
131848446 | |
[Zr+4].CC(C)O.CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-] |
C15H36O5Zr | |
MFCD00075157,MFCD00078045 | |
isopropanoltetrakis isopropanolato zirconium | |
propan-2-ol;propan-2-olate;zirconium(4+) |
Specifications
(decomposition) | |
Crystalline | |
MFCD00075157,MFCD00078045 | |
isopropanoltetrakis isopropanolato zirconium | |
NIJDLRJGRCHJDB-UHFFFAOYSA-N | |
propan-2-ol;propan-2-olate;zirconium(4+) | |
131848446 | |
Moisture sensitive |
14717-56-7 | |
C15H36O5Zr | |
UN3181 | |
Soluble in hot isopropanol,warm hexane and toluene; Tends to precipitate over time. | |
[Zr+4].CC(C)O.CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-] | |
387.67 | |
387.67 | |
Zirconium (IV) isopropoxide isopropanol complex |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 218-522-4
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only