missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, 99.7% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals

For vacuum deposition

98.89 € - 381.90 €

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11323408
SDS View Documents
Thermo Scientific Alfa Aesar
036319.18
50 g
98.89 €
50g
Please to purchase this item. Need a web account? Register with us today!
11333408
SDS View Documents
Thermo Scientific Alfa Aesar
036319.30
250 g
381.90 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For vacuum deposition. Raw material for the synthesis. Used as Additive of special glass, ceramics. It is also used as a detector of alpha-rays, which render it luminescent. As vacuum coating. Piezoelectric ceramics include filter ,speaker, the ultrasonic underwater acoustic detectors and so on . Stabilized oxide For refractory furnace linings, and in crucibles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-23-4
123.22
MCMNRKCIXSYSNV-UHFFFAOYSA-N
62395
O=[Zr]=O
O2Zr
MFCD00011310 MFCD01868884
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
dioxozirconium

Specifications

ca 5000°C
1314-23-4
Sintered Lump
O2Zr
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
MCMNRKCIXSYSNV-UHFFFAOYSA-N
dioxozirconium
62395
Odorless
Zirconium(IV) oxide, Hf < 75ppm
∽2700°C
White
99.7% metal basis excluding Hf
MFCD00011310 MFCD01868884
Insoluble in water. Soluble in hot and concentrated hydrofluoric acid and sulfuric acid. Dissolves on heating in a mixture of 2 parts H2SO4 and 1 part water.
O=[Zr]=O
123.22
123.22
5.89
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-227-2

missing translation for 'rtecsNumber' : ZH8800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.