missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, 99+% (metals basis excluding Hf), HfO{2} 2%, Thermo Scientific Chemicals

90.65 € - 1119.57 €

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381189
View Documents
Thermo Scientific Alfa Aesar
011395.30
250 g
90.65 €
250g
Please to purchase this item. Need a web account? Register with us today!
11371189
View Documents
Thermo Scientific Alfa Aesar
011395.A1
1 kg
247.23 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11391189
View Documents
Thermo Scientific Alfa Aesar
011395.A7
5 kg
1119.57 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zirconium(IV) oxide is used in the production of piezoelectric crystals and also used in the high frequency induction coils and ceramic glazes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-23-4
123.22
MCMNRKCIXSYSNV-UHFFFAOYSA-N
62395
O=[Zr]=O
O2Zr
MFCD00011310 MFCD01868884
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
dioxozirconium

Specifications

ca 5000°C
1314-23-4
≥99%, metal basis
MFCD00011310 MFCD01868884
Insoluble in water. Soluble in sulfuric acid and hydrofluoric acid.
O=[Zr]=O
123.22
123.22
Zirconium(IV) Oxide
∽2700°C
Powder
O2Zr
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
MCMNRKCIXSYSNV-UHFFFAOYSA-N
dioxozirconium
62395
5.89
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-227-2

missing translation for 'rtecsNumber' : ZH8800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only