Learn More
Zirconium(IV) oxide, calcia stabilized, 99.4% (metals basis excluding Hf), Thermo Scientific Chemicals
The main use of zirconia is in the production of ceramics. Zirconium dioxide is also used as the solid electrolyte in electrochromic devices As protective coating on particles of titanium dioxide pigments, as a refractory material, in insulation, abrasives and enamels.
44.32 € - 319.59 €
Chemical Identifiers
CAS | 1314-23-4 |
---|---|
Molecular Formula | O2Zr |
Molecular Weight (g/mol) | 123.22 |
MDL Number | MFCD00011310 MFCD01868884 |
InChI Key | MCMNRKCIXSYSNV-UHFFFAOYSA-N |
Synonym | zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler |
PubChem CID | 62395 |
IUPAC Name | dioxozirconium |
SMILES | O=[Zr]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348418
|
Thermo Scientific Alfa Aesar
040452.18 |
50 g |
44.32 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358418
|
Thermo Scientific Alfa Aesar
040452.30 |
250 g | N/A | ||||||
11368418
|
Thermo Scientific Alfa Aesar
040452.A1 |
1 kg |
319.59 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The main use of zirconia is in the production of ceramics. Zirconium dioxide is also used as the solid electrolyte in electrochromic devices As protective coating on particles of titanium dioxide pigments, as a refractory material, in insulation, abrasives and enamels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-23-4 | |
123.22 | |
MCMNRKCIXSYSNV-UHFFFAOYSA-N | |
62395 | |
O=[Zr]=O |
O2Zr | |
MFCD00011310 MFCD01868884 | |
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler | |
dioxozirconium |
Specifications
≈5000°C | |
1314-23-4 | |
O2Zr | |
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler | |
MCMNRKCIXSYSNV-UHFFFAOYSA-N | |
dioxozirconium | |
62395 | |
99.4% | |
Zirconium(IV) oxide |
∽2700°C | |
Powder | |
MFCD00011310 MFCD01868884 | |
Insoluble in water. soluble in HF,and hot Sulphuric acid. | |
O=[Zr]=O | |
123.22 | |
123.22 | |
Plastic bottle |
Safety and Handling
missing translation for 'einecsNumber' : 215-227-2
missing translation for 'rtecsNumber' : ZH8800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.