missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium(IV) oxide, cement, Thermo Scientific Chemicals
One component adhesive. Bonds to Zirconium oxide and Silicon. Exhibit good dielectric properties
108.54 € - 1036.00 €
Chemical Identifiers
CAS | 1314-23-4 |
---|---|
Molecular Formula | O2Zr |
Molecular Weight (g/mol) | 123.22 |
MDL Number | MFCD00011310 MFCD01868884 |
InChI Key | MCMNRKCIXSYSNV-UHFFFAOYSA-N |
Synonym | zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler |
PubChem CID | 62395 |
IUPAC Name | dioxozirconium |
SMILES | O=[Zr]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329028
|
Thermo Scientific Alfa Aesar
042493.30 |
250 g |
108.54 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11309028
|
Thermo Scientific Alfa Aesar
042493.A1 |
1 kg |
344.72 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319028
|
Thermo Scientific Alfa Aesar
042493.A7 |
5 kg | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-23-4 | |
123.22 | |
MCMNRKCIXSYSNV-UHFFFAOYSA-N | |
62395 | |
O=[Zr]=O |
O2Zr | |
MFCD00011310 MFCD01868884 | |
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler | |
dioxozirconium |
Specifications
1314-23-4 | |
Liquid, Paste | |
MFCD00011310 MFCD01868884 | |
MCMNRKCIXSYSNV-UHFFFAOYSA-N | |
dioxozirconium | |
62395 | |
Light brown liquid/paste |
Brown | |
O2Zr | |
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler | |
O=[Zr]=O | |
123.22 | |
Odorless | |
Zirconium(IV) oxide |
Safety and Handling
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
missing translation for 'einecsNumber' : 215-227-2
missing translation for 'rtecsNumber' : ZH8800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only