missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, Puratronic™, 99.978% (metals basis), Thermo Scientific Chemicals

233.00 € - 2092.00 €

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11306168
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011141.14
25 g
233.00 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11396158
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011141.22
100 g
701.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11386158
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011141.36
500 g
2092.00 €
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zirconium(IV) oxide is used as an additive in the production of piezoelectric crystals, in high-frequency induction coils and in ceramic glazes. It finds application as a solid electrolyte in electrochromic devices and precursor to the electroceramic lead zirconate titanate (PZT), as an insulator in transistors, as a thermal barrier coating in jet and diesel engines and as a diamond simulant in jewelry. It is used in oxygen sensors, fuel cell membranes and in orthopaedic implants of hip joints. It is coated on the silica gel surface and found to be used in the extraction of chromium from aqueous solutions at a pH of about 3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-23-4
123.22
MCMNRKCIXSYSNV-UHFFFAOYSA-N
62395
O=[Zr]=O
O2Zr
MFCD00011310 MFCD01868884
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
dioxozirconium

Specifications

∼5000°C
1314-23-4
Powder
ZrO2
14,10180
Soluble in alcohols,hydrofluoric and hot sulfuric acid. Insoluble in water.
O=[Zr]=O
123.22
123.22
Puratronic™
5.89 g/mL
Zirconium(IV) oxide
∼2700°C
White
O2Zr
MFCD00011310 MFCD01868884
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
MCMNRKCIXSYSNV-UHFFFAOYSA-N
dioxozirconium
62395
99.978%
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-227-2

missing translation for 'rtecsNumber' : ZH8800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only