missing translation for 'onlineSavingsMsg'
Learn More

Zirconium n-butoxide, 80% w/w in 1-butanol, Thermo Scientific Chemicals

29.55 € - 235.17 €

Chemical Identifiers

CAS 1071-76-7
Molecular Formula C16H40O4Zr+4
Molecular Weight (g/mol) 387.716
MDL Number MFCD00048765
InChI Key ASUYWGMDIAPAKI-UHFFFAOYSA-N
Synonym 1-butanol, zirconium 4+ salt 4:1, 1-butanol, zirconium 4++ salt
PubChem CID 122130442
IUPAC Name butan-1-ol;zirconium(4+)
SMILES CCCCO.CCCCO.CCCCO.CCCCO.[Zr+4]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11470927
View Documents
Thermo Scientific Alfa Aesar
B23317.18
50 g
29.55 €
50g
Please to purchase this item. Need a web account? Register with us today!
11480927
View Documents
Thermo Scientific Alfa Aesar
B23317.30
250 g
75.58 €
250g
Please to purchase this item. Need a web account? Register with us today!
11460927
View Documents
Thermo Scientific Alfa Aesar
B23317.0B
1000 g
235.17 €
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, acids, bases, halogens, alkali metals, water/ moisture.
Specifications

Chemical Identifiers

1071-76-7
387.716
ASUYWGMDIAPAKI-UHFFFAOYSA-N
122130442
CCCCO.CCCCO.CCCCO.CCCCO.[Zr+4]
C16H40O4Zr+4
MFCD00048765
1-butanol, zirconium 4+ salt 4:1, 1-butanol, zirconium 4++ salt
butan-1-ol;zirconium(4+)

Specifications

1071-76-7
[CH3(CH2)3O]4Zr
UN1120
38°C (100°F)
Hydrolyzes in water.
CCCCO.CCCCO.CCCCO.CCCCO.[Zr+4]
387.716
383.68
Alcohol-like
Moisture sensitive
Zirconium n-butoxide
C16H40O4Zr+4
MFCD00048765
4132649
1-butanol, zirconium 4+ salt 4:1, 1-butanol, zirconium 4++ salt
ASUYWGMDIAPAKI-UHFFFAOYSA-N
butan-1-ol;zirconium(4+)
122130442
80% w/w in 1-butanol
1.466
1.07
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H226-H335-H336-H315
Causes serious eye damage.
Flammable liquid and vapour.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H318-H335-H336

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS

missing translation for 'einecsNumber' : 213-995-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.