missing translation for 'onlineSavingsMsg'
Learn More

Zirconium nitride, 99.5% (metals basis excluding Hf), Hf <3%, Thermo Scientific Chemicals

158.79 € - 584.91 €

Chemical Identifiers

CAS 25658-42-8
Molecular Formula NZr
Molecular Weight (g/mol) 105.23
MDL Number MFCD00049642
InChI Key ZVWKZXLXHLZXLS-UHFFFAOYSA-N
Synonym zirconium nitride, zirconium nitride zrn, zirconium nitride,-325 mesh, zirconium nitride trace metals basis excluding hf 25g, zirconium nitride, powder, pore size 1-2 mum trace metals basis
PubChem CID 94359
IUPAC Name azanylidynezirconium
SMILES N#[Zr]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326508
View Documents
Thermo Scientific Alfa Aesar
012138.14
25 g
158.79 €
25g
Please to purchase this item. Need a web account? Register with us today!
11336508
View Documents
Thermo Scientific Alfa Aesar
012138.22
100 g
584.91 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zirconium nitride is used in cermets, refractories, and laboratory crucibles. It also used for coating medical devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25658-42-8
105.23
ZVWKZXLXHLZXLS-UHFFFAOYSA-N
94359
N#[Zr]
NZr
MFCD00049642
zirconium nitride, zirconium nitride zrn, zirconium nitride,-325 mesh, zirconium nitride trace metals basis excluding hf 25g, zirconium nitride, powder, pore size 1-2 mum trace metals basis
azanylidynezirconium

Specifications

2980°C
Powder
NZr
zirconium nitride, zirconium nitride zrn, zirconium nitride,-325 mesh, zirconium nitride trace metals basis excluding hf 25g, zirconium nitride, powder, pore size 1-2 mum trace metals basis
ZVWKZXLXHLZXLS-UHFFFAOYSA-N
azanylidynezirconium
94359
7.09 g/mL
25658-42-8
99.5% (metals basis excluding Hf)
MFCD00049642
Soluble in hydrofluoric acid. Insoluble in water.
N#[Zr]
105.23
105.23
Zirconium nitride
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III

missing translation for 'einecsNumber' : 247-166-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only