missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Zirconium Oxide Grinding Balls

Product Code. 11964382 Shop All Retsch Products
Click to view available options
For Use With (Equipment):
Cell and Tissue Disruption with 200,400 mm
Centrifugal Ball Mill S 100
High Energy Ball Mill Emax
MM 200 and MM 301
MM 200 and MM 400
MM 400
Planetary Ball Mill PM 200
Diameter (Metric):
0.1 mm
0.5 mm
1 mm
2 mm
3 mm
5 mm
12 mm
15 mm
40 mm
Unit Size:
Each
Pack of 200
This item is not returnable. View return policy

Product Code. 11964382

Brand: RETSCH 32.368.0003

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Product Type Grinding Balls
Diameter (Metric) 0.5 mm
For Use With (Equipment) High Energy Ball Mill Emax
Material Zirconium Oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.