missing translation for 'onlineSavingsMsg'
Learn More

Zirconium rod, 12.7mm (0.5in) dia, annealed, 99.2+% (metals basis excluding Hf), Hf 4.5% max, Thermo Scientific Chemicals

Product Code. 11325948
Click to view available options
Quantity:
5 cm
30 cm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Quantity 5 cm
Percent Purity ≥99.2%
Odor Odorless
Weight ≈8.24g/cm
Form Rod
Assay (metals basis excluding Hf), Hf 4.5% max.
Chemical Name or Material Zirconium rod, 12.7mm (0.5 in.) dia., Annealed, Temper: soft

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'