missing translation for 'onlineSavingsMsg'
Learn More

Zirconium rod, 6.2mm (0.24in) dia, annealed, 99% (metals basis excluding Hf), Hf 4.5% max, Thermo Scientific Chemicals

Form: approx. 1.96g/cm, Temper: soft

54.17 € - 182.91 €

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]
View More Specs

Products 2
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
11345948
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
010443.BS
10 cm
54.17 €
10cm
Please to purchase this item. Need a web account? Register with us today!
11335948
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
010443.BW
50 cm
182.91 €
50cm
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation

Chemische Identifikatoren

7440-67-7
91.22
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
zirconium
Zr
MFCD00011303 MFCD00148878
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
CHEBI:33342
[Zr]

Spezifikation

Gray to Silver
Gray to silver rod
99%
MFCD00011303 MFCD00148878
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zirconium
23995
Odorless
Zirconium, Annealed
7440-67-7
6.506g/cm3
Zr
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
[Zr]
91.22
CHEBI:33342
Rod
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

missing translation for 'einecsNumber' : 231-176-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate

RUO – Research Use Only