missing translation for 'onlineSavingsMsg'
Learn More

Zirconium silicate, Thermo Scientific Chemicals

30.60 € - 286.00 €

Chemical Identifiers

CAS 10101-52-7
Molecular Formula O4SiZr
Molecular Weight (g/mol) 183.31
MDL Number MFCD00085353
InChI Key GFQYVLUOOAAOGM-UHFFFAOYSA-N
Synonym zirconium silicate, hyacinth, zircon, zircon zr sio4, zirconite, zircosil 15, standard sf 200, ultrox 500w, a-pax 45m, zirconium iv silicate
PubChem CID 61775
IUPAC Name zirconium(4+);silicate
SMILES [Zr+4].[O-][Si]([O-])([O-])[O-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11490510
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030477.36
500 g
30.60 €
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11357278
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030477.A3
2 kg
84.20 €
2kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15401177
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030477.A9
10 kg
286.00 €
10kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zirconium silicate is used for manufacturing refractory materials for applications where resistance to corrosion by alkali materials is required. It is also used in production of some ceramics, enamels, and ceramic glazes. In enamels and glazes it serves as an opacifier. It can be also present in some cements. Another use of zirconium silicate is as beads for milling and grinding. Thin films of zirconium silicate and hafnium silicate produced by chemical vapor deposition. Potential alternative gate dielectric to silica.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10101-52-7
183.31
GFQYVLUOOAAOGM-UHFFFAOYSA-N
61775
[Zr+4].[O-][Si]([O-])([O-])[O-]
O4SiZr
MFCD00085353
zirconium silicate, hyacinth, zircon, zircon zr sio4, zirconite, zircosil 15, standard sf 200, ultrox 500w, a-pax 45m, zirconium iv silicate
zirconium(4+);silicate

Specifications

2550°C
White
O4SiZr
14,10181
Insoluble in water,acids,alkali and aqua regia.
[Zr+4].[O-][Si]([O-])([O-])[O-]
183.31
183.3
4.56 g/mL
10101-52-7
Powder
MFCD00085353
zirconium silicate, hyacinth, zircon, zircon zr sio4, zirconite, zircosil 15, standard sf 200, ultrox 500w, a-pax 45m, zirconium iv silicate
GFQYVLUOOAAOGM-UHFFFAOYSA-N
zirconium(4+);silicate
61775
Odorless
Zirconium silicate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 233-252-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only