Learn More
Zirconium silicide, 99.5% (metals basis excluding Hf), Hf 4% max, Thermo Scientific Chemicals
Description
Zirconium silicate is used for manufacturing refractory materials. It is also used in production of some ceramics, enamels, and ceramic glazes. In enamels and glazes it serves as an opacifier. zirconium silicate is as beads for milling and grinding.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Spezifikation
Color | Gray |
Physical Form | Powder |
Quantity | 25 g |
Solubility Information | Insoluble in water. It is insoluble in acids,alkali and aqua regia. |
Formula Weight | 147.39 |
Percent Purity | 99.5% |
Odor | Odorless |
Density | 4.88 g/mL |
Assay | (metals basis excluding Hf), Hf 4% |
Chemical Name or Material | Zirconium silicide |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.