missing translation for 'onlineSavingsMsg'
Learn More

Zirconium slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.5% (metals basis excluding Hf), Hf nominal 3%, Thermo Scientific Chemicals

55.68 € - 207.03 €

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11359068
View Documents
Thermo Scientific Alfa Aesar
042511.09
10 g
55.68 €
10g
Please to purchase this item. Need a web account? Register with us today!
11349068
View Documents
Thermo Scientific Alfa Aesar
042511.18
50 g
207.03 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-67-7
91.22
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
zirconium
Zr
MFCD00011303 MFCD00148878
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
CHEBI:33342
[Zr]

Specifications

7440-67-7
MFCD00011303 MFCD00148878
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zirconium
23995
99.5%
Odorless
Slug
Zirconium slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length
Zr
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
[Zr]
91.22
CHEBI:33342
Hf nominal 3%
≈0.16g/slug
(metals basis excluding Hf), Hf nominal 3%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-176-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only