missing translation for 'onlineSavingsMsg'
Learn More
Zirconium slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.5% (metals basis excluding Hf), Hf nominal 3%, Thermo Scientific Chemicals Product Code.: 11339068

Zirconium slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.5% (metals basis excluding Hf), Hf nominal 3%, Thermo Scientific Chemicals

Product Code. 11339068
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11339068

Brand: Thermo Scientific Alfa Aesar 042510.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Quantity 50 g
Percent Purity 99.5%
Concentration or Composition (by Analyte or Components) Hf nominal 3%
Odor Odorless
Weight ≈0.33g/slug
Form Slug
Assay (metals basis excluding Hf), Hf nominal 3%
Chemical Name or Material Zirconium slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberG06K22 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042510.09,042510.18,042511.18
Lot NumberU21F029 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042510.09,042510.18,042511.18
Lot NumberB20S004 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number042510.09,042510.18,042511.18
Lot NumberB17U024 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number042510.09,042510.18,042511.18
Lot NumberK18M26 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number042510.09,042510.18,042511.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.5% (metals basis excluding Hf), Hf nominal 3%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.