missing translation for 'onlineSavingsMsg'
Learn More

Zirconium sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.5% (metals basis excluding Hf), Thermo Scientific™

Product Code. 15414875
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-67-7
Molecular Formula Zr
MDL Number MFCD00011303 MFCD00148878
Quantity 1 Ea.
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
SMILES [Zr]
IUPAC Name zirconium
Molecular Weight (g/mol) 91.22
PubChem CID 23995
ChEBI CHEBI:33342
Percent Purity 99.5%
Odor Odorless
Assay (metals basis excluding Hf)
Chemical Name or Material Zirconium sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'