missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF259 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15558-1-AP-20UL
This item is not returnable.
View return policy
Description
The MW of this protein is 51 kDa, and this antibody specifically recognizes the 51 kDa protein.
ZNF259, also named as Zpr1, is an evolutionarily conserved protein characterized by two C4 zinc fingers and two Zpr1-specific conserved homology domains. Zpr1 was first identified in mammalian cells as a cytoplasmic protein that was capable of binding the intracellular domain of the inactivated EGF receptor (EGFR).Specifications
ZNF259 | |
Polyclonal | |
Unconjugated | |
ZPR1 | |
zinc finger protein 259, Zinc finger protein ZPR1, ZNF259, ZPR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8882 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75312 | |
ZPR1 | |
ZNF259 Fusion Protein Ag7980 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction