missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF600 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20100-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to ZNF600.
Consisting of around 63million baseswith over 1,400 genes,chromosome 19 makes up over 2% of human genomic DNA. Chromosome 19 includes a diversity of interesting genes and isrecognized for having the greatest gene density of the human chromosomes. It isthe genetic home for a number of immunoglobulin superfamily members including the killercell and leukocyte Ig-like receptors, a number of ICAMs, the CEACAMand PSG family, and Fcalpha receptors. Key genesfor eye color and haircolor also map to chromosome 19. Peutz-Jegherssyndrome,spinocerebellar ataxia type 6, the stroke disorder CADASIL, hypercholesterolemia and Insulin-dependent diabetes have been linked to chromosome 19. Translocations with chromosome 19 and chromosome 14 can be seen in some lymphoproliferative disorders and typicallyinvolve the proto-oncogene Bcl-3.Specifications
ZNF600 | |
Polyclonal | |
Unconjugated | |
ZNF600 | |
KR ZNF1, zinc finger protein 600, ZNF600 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
162966, 667666 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6ZNG1 | |
Zfp600, ZNF600 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction