Learn More
Zofenopril calcium, MedChemExpress
MedChemExpress Zofenopril Calcium (SQ26991) is an antioxidant that acts as an angiotensin-converting enzyme inhibitor.
Brand:  Medchem Express HY-B0655
Encompass_Preferred
 View more versions of this product
Product Code. 16428568
  • 172.80 € / 10mg
Explore more special offers
Description
Description
Zofenopril Calcium (SQ26991) is an antioxidant that acts as an angiotensin-converting enzyme inhibitor.
Specifications
Specifications
Zofenopril calcium
White
C22H22Ca0.5NO4S2
SQ26991
O=C(N(C[C@@H](SC1=CC=CC=C1)C2)[C@@H]2C([O-])=O)[C@H](C)CSC(C3=CC=CC=C3)=O.[0.5Ca2+]
448.58
Research
Neuroscience-Neurodegeneration
81938-43-4
Research
10 mg
DMSO : 2 mg/mL (4.46 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
448.58
99.56%
Solid