Learn More
Zofenopril calcium, MedChemExpress
MedChemExpress Zofenopril Calcium (SQ26991) is an antioxidant that acts as an angiotensin-converting enzyme inhibitor.
Brand: Medchem Express HY-B0655
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item is not returnable.
View return policy
Description
Zofenopril Calcium (SQ26991) is an antioxidant that acts as an angiotensin-converting enzyme inhibitor.Specifications
Zofenopril calcium | |
White | |
C22H22Ca0.5NO4S2 | |
SQ26991 | |
O=C(N(C[C@@H](SC1=CC=CC=C1)C2)[C@@H]2C([O-])=O)[C@H](C)CSC(C3=CC=CC=C3)=O.[0.5Ca2+] | |
448.58 | |
Research | |
Neuroscience-Neurodegeneration |
81938-43-4 | |
Research | |
50 mg | |
DMSO : 2 mg/mL (4.46 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) | |
448.58 | |
99.56% | |
Solid |