Learn More
Buerkle™ Zone Sampler Spiralus
Buerkle™ Zone Sampler Spiralus provides an excellent all-layer sample.
733.00 € - 918.00 €
Specifications
Product Type Zone Sampler Spiralus
Material Aluminum Anodised, PA
Includes 8 openings
Diameter (Metric) Outer 38 mm
View More Specs
Products 2
Product Code Brand Capacity (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Length (Metric) Price Quantity & Availability  
10532138
View Documents
Buerkle™
5318-0145
770 mL 145 cm
733.00 €
Each
In Stock
to see stock.
Add to basket
11857251
View Documents
Buerkle™
5318-0200
1100 mL 200 cm
918.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Representative sample of the container layers.
  • aluminum, anodised
  • Tip PA
  • Low weight
Specifications
Specifications
Zone Sampler Spiralus
Aluminum Anodised, PA
8 openings
38 mm