Learn More
Invitrogen™ ZOOM™ IPGRunner™ Cassette
For use with the ZOOM IPGRunner Mini-Cell
Brand: Invitrogen™ ZM0003
Includes: Electrode wicks and sealing tape.
Description
The ZOOM IPGRunner Cassette provides a convenient way to rehydrate 7.0cm ZOOM Strips and perform isoelectric focusing of up to six 7.0cm ZOOM Strips (IPG strips) without requiring separate units for rehydration and isoelectric focusing.
2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Specifications
• Zoom IPGRunner Cassettes (10) • Sealing tape (22) • Electrode wicks (22) |
|
IPGRunner, ZOOM | |
Room Temperature |
ZOOM IPGRunner System | |
10 cassette(s) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.