Learn More
Invitrogen™ ZOOM™ IPGRunner™ Combo Kit
Oil-free, 2D electrophoresis in a minigel format
Brand: Invitrogen™ ZM0002
1657.85 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Includes: ZOOM IPGRunner Mini-Cell, electrode wicks, sealing tape, 10 ZOOM IPGRunner Cassettes, and 12 ZOOM Strips, pH 3-10 NL
Description
The ZOOM IPGRunner System typically completes first dimension separation isoelectric focusing (IEF) in less than three hours. The minigel design of the system is easy to handle, eliminates mineral oil overlays, and allows processing of up to 12 samples at once for high-throughput usage.
- The ZOOM IPGRunner Combo Kit includes everything needed for first dimension separation: ZOOM IPGRunner Mini-Cell; 10 ZOOM IPGRunner Cassettes; 12 ZOOM Strips, pH 3-10NL
- For second dimension separation, Novex™ ZOOM gels and the XCell SureLock™ Mini-Cell are recommended (both are available separately)
ZOOM IPGRunner Mini-Cell
- Consists of a ZOOM IPGRunner Core, a mini-cell chamber, and a lid
- Can be used to perform isoelectric focusing of up to 12 IPG strips (ZOOM Strips) loaded in two ZOOM IPGRunner Cassettes
- Used to perform oil-free rehydration and isoelectric focusing of up to six 7.0cm IPG strips (ZOOM Strips) without requiring separate units for rehydration and isoelectric focusing
- Disposable
- 7cm long
- Contain a thin layer of polyacrylamide gel with a fixed pH gradient
- Clearly labeled with a unique identifying number, pH range, and orientation marks
- Supplied attached to a tri-fold card for easy access and removal
- ZOOM Gels are precast polyacrylamide gels designed for 2D analysis of proteins following isoelectric focusing with ZOOM Strips
- They are recommended for use with our XCell SureLock™ Mini-Cell
- ZOOM Gels contain an IPG well and a molecular weight marker well; the IPG well is designed to accommodate the 7.0cm ZOOM Strips
- Two types of ZOOM Gels are available: NuPAGE™ Novex 4–12% Bis-Tris ZOOM Gel and Novex 4–20% Tris-Glycine ZOOM Gel
2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Specifications
The ZOOM™ IPGRunner™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini-Cell chamber The ZOOM™ IPGRunner™ Combo Kit includes the ZOOM™ IPGRunner™ Mini-Cell, electrode wicks, sealing tape, 10 ZOOM™ IPGRunner™ Cassettes, and 12 ZOOM™ Strips, pH 3-10 NL The ZOOM™ IPGRunner™ Retrofit Kit for XCell SureLock™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid. The ZOOM™ IPGRunner™ Cassettes include electrode wicks and sealing tape |
|
ZOOM™ IPG Strips | |
IPGRunner, ZOOM |
XCell SureLock™ Mini, ZOOM™ IPGRunner™ Mini | |
7 cm Strips | |
1 kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.