missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 100 μL

Clear, autoclavable tips; color-coded racks.

331.71 € - 869.41 €

Specifications

Volume (Metric) 100 μL
Color Clear
Rack Color Gray
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
View More Specs

Products 5
Product Code Brand Tip Style Sterility Filtered Description Price Quantity & Availability  
Product Code Brand Tip Style Sterility Filtered Description Price Quantity & Availability  
12726769
View Documents
Axygen™
ZTF-100-L-R-S
Automation, Round Sterile Filtered Maxymum Recovery
869.41 €
Pack of 4800
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
12786669
View Documents
Axygen™
ZT-100-R
Automation, Round Non-sterile Non-filtered Standard
353.10 €
Pack of 4800
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
 
11346064
View Documents
Axygen™
ZT-100-L-R
Automation, Round Non-sterile Non-filtered Maxymum Recovery This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12736769
View Documents
Axygen™
ZTF-100-R-S
Automation, Round Sterile Filtered Standard
782.44 €
Pack of 4800
In Stock
to see stock.
Add to basket
 
12776669
View Documents
Axygen™
ZT-100-L-R-S
Automation, Round Sterile Non-filtered Maxymum Recovery This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Tip format: 96 tip and 384 tip
  • Tip volume range: 25 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: HV96, CAS-4200-1 (20 - 150 μL/sec.), LV96, LV384
  • Rack format: Non-ANSI (96) and ANSI/SLAS (384)
  • Rack material: Polypropylene

Order Info

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

Specifications

Specifications

100 μL
Gray
Non-ANSI
Automation, Round
Caliper Zymark
Clear
Autoclavable
Rack
Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.