missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Zymolyase™
For lysing the cell walls of viable yeast cells and preparing yeast spheroblasts
Brand: MP Biomedicals™ 08320921
725.81 EUR valid until 2025-12-31
Use promo code "24274" to get your promotional price.
This item is not returnable.
View return policy
Description
- Highly purified preparation from Arthrobacter luteus
- Efficiently hydrolyzes poly (β-1,3 glucose)-like yeast cell-wall glucan
- One unit is defined as the amount of activity causing a decrease in A800 of 30%
- Available with lytic activity of 20,000U/g (Zymolyase 20T) or 100,000U/g (Zymolyase 100T)
Specifications
20,000 U/mg | |
Used to lyse living yeast cell walls to produce protoplast or spheroplast of various strains of Yeast cells | |
Arthrobacter luteus | |
Zymolyase 20T |
5 to 10 | |
2°C to 8°C | |
1 g | |
Lyophilized Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- ZYMOLYASE 20T
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction