missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Microcheck™ Beverage Monitor

Easy to use, versatile, bulk packaged disposable filter funnels are ideal for microbial analysis.

Brand:  Cytiva (formerly Pall Lab) 4761

Additional Details : Weight : 1.59600kg

 View more versions of this product

Product Code. 17104401

  • 182.00 € / Pack of 50
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

  • Can convert to a petri dish for culturing or easily remove the membrane for placing on separate agar dish
  • Product not approved for use in medical clinical surgical or other patient applications
Specifications

Specifications

100 mL
Polystyrene Cover, Polypropylene Cylinder/Base, Polyethylene Adapter Material, Cellulose Support Pad
50/Pk.
GN-6 Metricel Mixed Cellulose Ester
Black
0.45 μm
Gamma Irradiated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers