missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Microcheck™ Beverage Monitor
Easy to use, versatile, bulk packaged disposable filter funnels are ideal for microbial analysis.
182.00 €
Specifications
Capacity (Metric) | 100 mL |
---|---|
Color | Black |
Description | Polystyrene Cover, Polypropylene Cylinder/Base, Polyethylene Adapter Material, Cellulose Support Pad |
Pore Size | 0.45 μm |
Quantity | 50/Pk. |
Product Code | Brand | Material (Membrane) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material (Membrane) | Price | Quantity & Availability | |||||
17104401
|
Cytiva (formerly Pall Lab)
4761 |
GN-6 Metricel Mixed Cellulose Ester |
182.00 €
Pack of 50 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
17124401
|
Cytiva (formerly Pall Lab)
4763 |
Metricel Black Membrane Modified PES |
182.00 €
Pack of 50 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
Description
- Can convert to a petri dish for culturing or easily remove the membrane for placing on separate agar dish
- Product not approved for use in medical clinical surgical or other patient applications
Specifications
100 mL | |
Polystyrene Cover, Polypropylene Cylinder/Base, Polyethylene Adapter Material, Cellulose Support Pad | |
50/Pk. |
Black | |
0.45 μm | |
Gamma Irradiated |