Learn More
Sartorius Minisart™ NML Syringe Filters, Sterile
Provides fast and reliable filtration of small to medium volumes, complete with ready-to-connect filter units. Sartorius Minisart™ NML Syringe Filters, Sterile eliminate time consuming preparatory work, such as changing out the filter and cleaning the filtration system.
155.00 € - 227.00 €
Specifications
Disposable | Single-use |
---|---|
Material | Surfactant-free Cellulose Acetate |
Filtration Area | 6.2 cm2 |
For Use With (Application) | Preparation of aqueous liquids |
Hold up Volume (After Air Purge) | 0.15 mL |
Product Code | Brand | Pore Size | Color | Sterilization Method | Outlet Style | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Color | Sterilization Method | Outlet Style | Price | Quantity & Availability | ||
11795745
|
Sartorius
17528----------K |
0.2 μm | Pink | Male Luer Lock |
179.00 €
Pack of 50 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
10509821
|
Sartorius
16534----------K |
0.2 μm | Blue | ETO-sterile | Male Luer Lock |
227.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
10076891
|
Sartorius
16534--------GUK |
0.2 μm | Blue | Gamma-sterile, individually packaged | Male Luer Lock |
179.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
10527401
|
Sartorius
17597----------K |
0.2 μm | Blue | ETO-sterile | Male Luer Slip |
189.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
11736035
|
Sartorius
16569----------K |
0.65 μm | Pink | ETO-sterile | Male Luer Lock |
161.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
10009230
|
Sartorius
17598----------K |
0.45 μm | Yellow | ETO-sterile | Male Luer Slip |
185.76 €
Pack of 50 |
In Stock
Log in to see stock. |
||
11786953
|
Sartorius
16555--------GUK |
0.45 μm | Yellow | Gamma-sterile, individually packaged | Male Luer Lock |
188.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
10109180
|
Sartorius
16555----------K |
0.45 μm | Yellow | ETO-sterile | Male Luer Lock |
179.00 €
Pack of 50 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||
10413131
|
Sartorius
16592----------K |
0.8 μm | Green | ETO-sterile | Male Luer Lock |
155.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
11746035
|
Sartorius
16592--------GUK |
0.8 μm | Green | Gamma-sterile, individually packaged | Male Luer Lock |
155.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
10189220
|
Sartorius
17593----------K |
1.2 μm | Red | ETO-sterile | Male Luer Lock |
197.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
10230891
|
Sartorius
17594----------K |
5 μm | Brown | ETO-sterile | Male Luer Lock |
180.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
Description
Provides fast and reliable filtration of small to medium volumes, complete with ready-to-connect filter units. Sartorius Minisart™ NML Syringe Filters, Sterile eliminate time consuming preparatory work, such as changing out the filter and cleaning the filtration system.
Fast and reliable filtration of small to medium volumes, complete with ready-to-connect filter units eliminate time consuming preparatory work, such as changing out the filter and cleaning the filtration system. These labor saving units are used for a wide variety of applications: sample cleaning, particle removal, and sterile filtration.
- Handy and readily available
- Easy to use
- Proven quality
- Economical
- Available in a wide array of different types
- 100% tested during manufacture
Specifications
Single-use | |
6.2 cm2 | |
0.15 mL | |
Female Luer Lock |
Surfactant-free Cellulose Acetate | |
Preparation of aqueous liquids | |
Meta acrylate butadiene styrene polymerisate (MBS) | |
28 mm |