man

Test

CAS RN 10025-97-5

ClClClClIr

CAS RN 10025-97-5

IUPAC Name: iridium(4+) tetrachloride
Synonyms: iridium(4+) tetrachloride
Molecular Weight (g/mol): 334.02
Molecular Formula: Cl4Ir
InChi Key: CALMYRPSSNRCFD-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Ir+4]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)