accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1003-21-0

CAS RN 1003-21-0

IUPAC Name: 5-bromo-1-methyl-1H-imidazole
Synonyms: 5-bromo-1-methylimidazole
Molecular Weight (g/mol): 161
Molecular Formula: C4H5BrN2
InChi Key: HATLLUIOEIXWGD-UHFFFAOYSA-N
SMILES: CN1C=NC=C1Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.