accessibility menu, dialog, popup

Test

CAS RN 1003-73-2

CH3NHOHC

CAS RN 1003-73-2

IUPAC Name: 3-methyl-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate
Synonyms: 3-methyl-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate
Molecular Weight (g/mol): 110.14
Molecular Formula: C6H8NO
InChi Key: FHJYMRFPLWFYPD-UHFFFAOYNA-N
SMILES: CC1=CC=C[NH++]([O-])[CH-]1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)