accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10031-90-0

CAS RN 10031-90-0

IUPAC Name: ethyl 3-(furan-2-yl)propanoate
Synonyms: ethyl 3-(furan-2-yl)propanoate
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: OWIWZQQFSTZZIG-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.