man

Test

CAS RN 10039-56-2

H2ONaOPHOH

CAS RN 10039-56-2

IUPAC Name: sodium hydrate oxophosphinite
Synonyms: sodium hypophosphite
Molecular Weight (g/mol): 103.98
Molecular Formula: H2NaO3P
InChi Key: ACUGTEHQOFWBES-UHFFFAOYSA-M
SMILES: O.[Na+].[O-]P=O
Quantity 
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (4)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (4)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (3)
  • (4)