accessibility menu, dialog, popup

Test

CAS RN 1005-28-3

CH3NH3CNSO

CAS RN 1005-28-3

IUPAC Name: 2-(dimethylamino)-1,3-thiazole-5-carbaldehyde
Synonyms: 2-(dimethylamino)-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 156.2
Molecular Formula: C6H8N2OS
InChi Key: ZOPJEHPVWLTDJW-UHFFFAOYSA-N
SMILES: CN(C)C1=NC=C(S1)C=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results