accessibility menu, dialog, popup

Test

CAS RN 1006-68-4

ON

CAS RN 1006-68-4

IUPAC Name: 5-phenyl-1,3-oxazole
Synonyms: 5-phenyl-1,3-oxazole
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: YPYPBEGIASEWKA-UHFFFAOYSA-N
SMILES: O1C=NC=C1C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)