accessibility menu, dialog, popup

Test

CAS RN 1016-78-0

ClO

CAS RN 1016-78-0

IUPAC Name: (3-chlorophenyl)(phenyl)methanone
Synonyms: (3-chlorophenyl)(phenyl)methanone
Molecular Weight (g/mol): 216.66
Molecular Formula: C13H9ClO
InChi Key: CPLWKNRPZVNELG-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)C(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results