missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 0.5 mL Conical Screw Cap Tubes: Presterilized
Features chemical resistant O-ring
259.31 € - 1895.80 €
Specifications
Certifications/Compliance | RNase, DNase, pyrogen-free |
---|---|
Closure Material | Polypropylene |
Capacity (Metric) | 0.5 mL |
Material | Polypropylene |
Format | Non-looped |
Product Code | Brand | Closure Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Color | Price | Quantity & Availability | ||||||
12769488
|
Axygen™
SCT-050-A-S |
Assorted | N/A | N/A | N/A | |||||
11330345
|
Axygen™
SCT-050-A-S |
Assorted |
259.31 €
Pack of 500 |
Estimated Shipment: 04-09-2025
Log in to see stock. |
||||||
12709498
|
Axygen™
SCT-050-C-S |
Clear | N/A | N/A | N/A | |||||
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
- Presterilized cap tubes have non-looped caps attached with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
- Available in range of colors for easy sample identification
Specifications
RNase, DNase, pyrogen-free | |
0.5 mL | |
Non-looped | |
Sterile | |
Conical |
Polypropylene | |
Polypropylene | |
O-ring | |
Screw Cap | |
20,000 x g |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ 0.5 mL Conical Screw Cap Tubes: Presterilized