missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 283°C to 287°C |
Color | White to Beige |
Boiling Point | 397°C to 400°C |
Flash Point | 380°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Plastic bottle |
Quantity | 2.5 kg |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0459176 | Certificate of Analysis | 31/01/2025 | 105320025,105325000,105321000 |
A0461360 | Certificate of Analysis | 11/06/2024 | 105320025,105325000,105321000 |
A0462570 | Certificate of Analysis | 16/05/2024 | 105320025,105325000,105321000 |
A0459176 | Certificate of Analysis | 08/01/2024 | 105320025,105325000,105321000 |
A0444449 | Certificate of Analysis | 25/08/2023 | 105320025,105325000,105321000 |
Safety and Handling


Product Identifier
- 1,2,4,5-Benzenetetracarboxylic anhydride
- Danger
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : 1,2,4,5-Benzenetetracarboxylic anhydride
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction