missing translation for 'onlineSavingsMsg'
Learn More
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals Product Code.: 10579630

1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals

Product Code. 10579630
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10579630

Brand: Thermo Scientific Acros 105325000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-32-7
Molecular Formula C10H2O6
Molecular Weight (g/mol) 218.12
MDL Number MFCD00005005
InChI Key ANSXAPJVJOKRDJ-UHFFFAOYSA-N
Synonym pyromellitic dianhydride, pyromellitic anhydride, pyromellitic acid anhydride, 1,2,4,5-benzenetetracarboxylic anhydride, pyromellitic acid dianhydride, 1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, pmda, benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetraone, unii-2z22t30qzq, 1,2,4,5-benzenetetracarboxylic dianhydrideShow MoreShow Less
PubChem CID 6966
IUPAC Name furo[3,4-f][2]benzofuran-1,3,5,7-tetrone
SMILES O=C1OC(=O)C2=CC3=C(C=C12)C(=O)OC3=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 283°C to 287°C
Color White to Beige
Boiling Point 397°C to 400°C
Flash Point 380°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Beilstein 19,196
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in acetone
Formula Weight 218.12
Percent Purity 99%
Physical Form Fine Powder
Chemical Name or Material 1,2,4,5-Benzenetetracarboxylic anhydride, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459176 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number105320025,105325000,105321000
Lot NumberA0461360 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number105320025,105325000,105321000
Lot NumberA0462570 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number105320025,105325000,105321000
Lot NumberA0459176 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number105320025,105325000,105321000
Lot NumberA0444449 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number105320025,105325000,105321000
compliance-icons compliance-icons
Product Identifier
  • 1,2,4,5-Benzenetetracarboxylic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P363-Wash contaminated clothing before reuse.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 1H,3H-Benzo[1,2-c:4,5-c']difuran-1,3,5,7-tetrone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.