Learn More
1,2,4-Triazole, 99%, Thermo Scientific Chemicals
Description
1,2,4-Triazole is widely used as an intermediate in the manufacture of pharmaceuticals. It acts as an anti-scaling agent and antifungal compound. It is widely used in antifungals such as fluconazole anditraconazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,2,4-Triazole is widely used as an intermediate in the manufacture of pharmaceuticals. It acts as an anti-scaling agent and antifungal compound. It is widely used in antifungals such as fluconazole anditraconazole.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and bases.

Specifications
Specifications
Melting Point | 118°C to 122°C |
Boiling Point | 260°C |
Flash Point | 158°C (316°F) |
Quantity | 25 g |
Beilstein | 104767 |
Merck Index | 14,9605 |
Solubility Information | Soluble in water. |
Formula Weight | 69.07 |
Percent Purity | 99% |
Chemical Name or Material | 1,2,4-Triazole |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z13E046 | Certificate of Analysis | 12/04/2022 | A11597.22,A11597.14,A11597.36 |
Y20A003 | Certificate of Analysis | 22/11/2021 | A11597.22,A11597.14,A11597.36 |
5001V01V | Certificate of Analysis | 22/11/2021 | A11597.22,A11597.14,A11597.36 |
I9712A | Certificate of Analysis | 22/11/2021 | A11597.22,A11597.14,A11597.36 |
S12B012 | Certificate of Analysis | 22/11/2021 | A11597.22,A11597.14,A11597.36 |
Safety and Handling


- 1,2,4-Triazole
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 1,2,4-Triazole
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.