missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Trichlorobenzene, ReagentPlus™, ≥99%, Honeywell™
Reagent Grade,≥99%
45.61 € - 517.74 €
Chemical Identifiers
CAS | 120-82-1 |
---|---|
Molecular Formula | C6H3Cl3 |
Molecular Weight (g/mol) | 181.44 |
MDL Number | MFCD00000547 |
InChI Key | PBKONEOXTCPAFI-UHFFFAOYSA-N |
Synonym | benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene |
PubChem CID | 13 |
ChEBI | CHEBI:28222 |
IUPAC Name | 1,2,4-trichlorobenzene |
SMILES | C1=CC(=C(C=C1Cl)Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15681270
|
Honeywell Research Chemicals
132047-1L |
1 L | Glass Bottle | ||||||
15691270
|
Honeywell Research Chemicals
132047-2.5L |
2.5 L | Glass Bottle |
134.45 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15601280
|
Honeywell Research Chemicals
132047-20L |
20 L | Stainless Steel Can | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
120-82-1 | |
181.44 | |
PBKONEOXTCPAFI-UHFFFAOYSA-N | |
13 | |
1,2,4-trichlorobenzene |
C6H3Cl3 | |
MFCD00000547 | |
benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene | |
CHEBI:28222 | |
C1=CC(=C(C=C1Cl)Cl)Cl |
Specifications
17°C | |
Colorless | |
213.5°C | |
110°C (230°F) | |
Liquid | |
C6H3Cl3 | |
1mmHg (40°C) | |
956819 | |
PBKONEOXTCPAFI-UHFFFAOYSA-N | |
1,2,4-trichlorobenzene | |
13 | |
181.45g/mol | |
ReagentPlus™ |
120-82-1 | |
1.454g/mL at 25°C | |
6.6% (150°F) | |
1060°F | |
n20/D 1.571(lit.) | |
MFCD00000547 | |
UN2321 | |
benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene | |
C1=CC(=C(C=C1Cl)Cl)Cl | |
181.44 | |
CHEBI:28222 | |
≥99% | |
1,2,4-Trichlorobenzene |
Safety and Handling
P301 + P312 + P330
H302-H315-H410
missing translation for 'dotInformation' : 6.1/PGIII
missing translation for 'einecsNumber' : 204-428-0
missing translation for 'rtecsNumber' : DC2100000