missing translation for 'onlineSavingsMsg'
Learn More
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene, 97+%, Thermo Scientific Chemicals Product Code.: 15475866

1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene, 97+%, Thermo Scientific Chemicals

Product Code. 15475866
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15475866

Brand: Thermo Scientific Alfa Aesar H60592.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as highly efficient privileged ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as highly efficient privileged ligands.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147253-67-6
Molecular Formula C18H28P2
Molecular Weight (g/mol) 306.37
MDL Number MFCD00142336
InChI Key AJNZWRKTWQLAJK-KLHDSHLOSA-N
Synonym r,r-me-duphos, methyl-duphos, r,r, r,r-methyl-duphos, unii-h5w03d1haq, h5w03d1haq, --1,2-bis 2r,5r-2,5-dimethylphospholano benzene, r,r-meduphos, methyl-duphos,-, 2r,5r-me-duphosShow MoreShow Less
PubChem CID 2734551
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane
SMILES CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C

Specifications

Melting Point 66°C to 76°C
Quantity 250 mg
Beilstein 4810602
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 306.36
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT18E017 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberH60592.03,H60592.MD
Lot NumberZ11J027 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog NumberH60592.03,H60592.MD
Lot NumberK13Y022 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60592.03,H60592.MD
Lot NumberZ15D026 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60592.03,H60592.MD
Lot NumberT18E017 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60592.03,H60592.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.